Assay Details
| Gene Symbol: MLLT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324296.1 | NP_001311225.1 | 1 - 2 | 488 | 69 | |
| NM_001324297.1 | NP_001311226.1 | 1 - 2 | 488 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001324296.1, NM_001324297.1 transcripts.
Set Membership: |
|

MLLT10
Genomic Map