Assay Details
| Gene Symbol: HKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026680.1 | XP_016882169.1 | 5 - 6 | 574 | 59 | |
| XM_017026681.1 | XP_016882170.1 | 4 - 5 | 522 | 59 | ||
| XM_017026682.1 | XP_016882171.1 | 5 - 6 | 574 | 59 | ||
| GenBank mRNA | AK128116.1 | - | 4 - 5 | 715 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017026680.1, XM_017026681.1, XM_017026682.1 transcripts.
Set Membership: |
|

HKR1
Genomic Map