Assay Details
| Gene Symbol: HKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026672.1 | XP_016882161.1 | 3 - 4 | 766 | 78 | |
| XM_017026674.1 | XP_016882163.1 | 7 - 8 | 1132 | 78 | ||
| XM_017026676.1 | XP_016882165.1 | 4 - 5 | 427 | 78 | ||
| XM_017026677.1 | XP_016882166.1 | 4 - 5 | 414 | 78 | ||
| XM_017026680.1 | XP_016882169.1 | 7 - 8 | 794 | 78 | ||
| XM_017026681.1 | XP_016882170.1 | 6 - 7 | 742 | 78 | ||
| XM_017026683.1 | XP_016882172.1 | 6 - 7 | 653 | 78 | ||
| XM_017026684.1 | XP_016882173.1 | 5 - 6 | 515 | 78 | ||
| XM_017026685.1 | XP_016882174.1 | 3 - 4 | 360 | 78 | ||
| XM_017026700.1 | XP_016882189.1 | 5 - 6 | 524 | 78 | ||
| GenBank mRNA | AK223616.1 | - | 4 - 5 | 418 | 78 | |
| AK299969.1 | - | 4 - 5 | 392 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017026700.1 transcripts.
Set Membership: |
|

HKR1
Genomic Map