Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182614.2 | NP_872420.1 | 5 - 6 | 451 | 88 | |
| XM_017020560.1 | XP_016876049.1 | 4 - 5 | 414 | 88 | ||
| GenBank mRNA | BC018995.2 | - | 5 - 6 | 451 | 4300679 | 88 |
More Information
Set Membership: |
|

TMEM255B
Genomic Map