Assay Details
| Gene Symbol: PTS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000317.2 | NP_000308.1 | 4 - 5 | 325 | 114 | |
| GenBank mRNA | AK310768.1 | - | 3 - 4 | 279 | 114 | |
| BC009686.1 | - | 4 - 5 | 289 | 3901665 | 114 | |
| D17400.1 | - | 4 - 5 | 314 | 114 | ||
| M97655.1 | - | 4 - 5 | 255 | 114 | ||
More Information
Set Membership: |
|

PTS
Genomic Map