Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003704.2 | NP_001003704.1 | 2 - 3 | 452 | 65 | |
| GenBank mRNA | BQ187590.1 | - | 2 - 3 | 234 | 65 | |
More Information
Set Membership: |
|

MTFP1
Genomic Map