Assay Details
| Gene Symbol: LAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006014.4 | NP_006005.2 | 1 - 2 | 528 | 108 | |
| GenBank mRNA | BC015744.1 | - | 1 - 2 | 528 | 4899044 | 108 |
| BC019012.2 | - | 1 - 2 | 150 | 108 | ||
| BC062330.1 | - | 1 - 2 | 241 | 5485603 | 108 | |
| BQ649305.1 | - | 1 - 2 | 195 | 108 | ||
| X92896.1 | - | 1 - 2 | 182 | 107 | ||
More Information
Set Membership: |
|

LAGE3
Genomic Map