Assay Details
| Gene Symbol: OXLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039842.2 | NP_001034931.1 | 1 - 2 | 127 | 109 | |
| GenBank mRNA | BC090923.1 | - | 1 - 2 | 78 | 30335041 | 109 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC090923.1, NM_001039842.2 transcripts.
Set Membership: |
|

OXLD1
Genomic Map