Assay Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042493.2 | NP_001035958.1 | 1 - 2 | 49 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001042493.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
