Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016644.2 | NP_057728.1 | 2 - 3 | 487 | 91 | |
| GenBank mRNA | AF242769.1 | - | 2 - 3 | 441 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR16| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016644.2 | NP_057728.1 | 2 - 3 | 487 | 91 | |
| GenBank mRNA | AF242769.1 | - | 2 - 3 | 441 | 91 | |
Set Membership: |
|