Assay Details
| Gene Symbol: LY6G5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021221.2 | NP_067044.2 | 1 - 2 | 63 | 94 | |
| GenBank mRNA | AJ315545.1 | - | 1 - 2 | 63 | 94 | |
| AK289364.1 | - | 1 - 2 | 847 | 94 | ||
| CF264683.1 | - | 2 - 3 | 96 | 94 | ||
| HE864450.1 | - | 1 - 2 | 83 | 94 | ||
More Information
Set Membership: |
|

LY6G5B
Genomic Map