Assay Details
| Gene Symbol: ADAM30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021794.3 | NP_068566.2 | 1 | 2225 | 72 | |
| GenBank mRNA | AF171932.1 | - | 1 | 2037 | 72 | |
| AF171933.1 | - | 1 | 2037 | 72 | ||
| AK292483.1 | - | 1 | 2216 | 72 | ||
| AY358734.1 | - | 1 | 2199 | 72 | ||
| BC028372.2 | - | 1 | 2196 | 4831258 | 72 | |
More Information
Set Membership: |
|

ADAM30
Genomic Map