Assay Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080748.2 | NP_542786.1 | 1 - 2 | 150 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_080748.2 transcripts.
Set Membership: |
|

ROMO1
Genomic Map