Assay Details
| Gene Symbol: NEURL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080749.3 | NP_542787.1 | 1 - 2 | 1039 | 55 | |
| GenBank mRNA | AJ295985.1 | - | 1 - 2 | 744 | 55 | |
| AK054821.1 | - | 1 - 2 | 1039 | 55 | ||
| AW515245.1 | - | 1 - 2 | 240 | 55 | ||
| BC074737.2 | - | 1 - 2 | 769 | 30915194 | 55 | |
| BC105935.1 | - | 1 - 2 | 929 | 40031539 | 55 | |
| BC107485.1 | - | 1 - 2 | 929 | 40031537 | 55 | |
| HY058356.1 | - | 1 - 2 | 408 | 55 | ||
More Information
Set Membership: |
|

NEURL2
Genomic Map