Assay Details
| Gene Symbol: TIGD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145702.2 | NP_663748.1 | 1 | 815 | 81 | |
| GenBank mRNA | AK056329.1 | - | 1 | 741 | 81 | |
| BC035143.1 | - | 1 | 820 | 5264460 | 81 | |
| BC063500.1 | - | 1 | 775 | 5248661 | 81 | |
More Information
Set Membership: |
|

TIGD1
Genomic Map