Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173549.2 | NP_775820.2 | 1 - 2 | 207 | 90 | |
| GenBank mRNA | AK096872.1 | - | 1 - 2 | 207 | 90 | |
| BC062359.1 | - | 1 - 2 | 151 | 5015294 | 90 | |
More Information
Set Membership: |
|

ERICH5
Genomic Map