Assay Details
| Gene Symbol: CMTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181272.2 | NP_851789.1 | 1 - 2 | 206 | 96 | |
| NM_181283.2 | NP_851800.1 | 1 - 2 | 206 | 96 | ||
| GenBank mRNA | AY174121.1 | - | 1 - 2 | 210 | 96 | |
| AY174122.1 | - | 1 - 2 | 210 | 96 | ||
More Information
Set Membership: |
|

CMTM1
Genomic Map