Assay Details
| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181840.1 | NP_862823.1 | 1 - 2 | 225 | 66 | |
| GenBank mRNA | AB087138.1 | - | 1 - 2 | 225 | 66 | |
More Information
Set Membership: |
|

KCNK18
Genomic Map