Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003958.3 | NP_003949.1 | 1 - 2 | 306 | 110 | |
| NM_183078.2 | NP_898901.1 | 1 - 2 | 306 | 110 | ||
| NR_046399.1 | 1 - 2 | 306 | 110 | |||
| XM_006715241.2 | XP_006715304.1 | 1 - 2 | 306 | 110 | ||
| XM_006715242.2 | XP_006715305.1 | 1 - 2 | 306 | 110 | ||
| XM_017011464.1 | XP_016866953.1 | 1 - 2 | 205 | 110 | ||
| GenBank mRNA | AB014546.1 | - | 1 - 2 | 225 | 110 | |
| AF334675.1 | - | 1 - 2 | 113 | 110 | ||
| AK222765.1 | - | 1 - 2 | 224 | 110 | ||
| AK298319.1 | - | 1 - 2 | 224 | 110 | ||
| AK308840.1 | - | 1 - 2 | 213 | 110 | ||
| AL711279.1 | - | 1 - 2 | 264 | 110 | ||
| BC007517.2 | - | 1 - 2 | 235 | 3028746 | 110 | |
| BT007446.1 | - | 1 - 2 | 113 | 110 | ||
| DA064222.1 | - | 1 - 2 | 306 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298319.1, XM_017011464.1 transcripts.
Set Membership: |
|

RNF8
Genomic Map