Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003958.3 | NP_003949.1 | 2 - 3 | 435 | 62 | |
| NM_183078.2 | NP_898901.1 | 2 - 3 | 435 | 62 | ||
| XM_006715241.2 | XP_006715304.1 | 2 - 3 | 435 | 62 | ||
| XM_006715242.2 | XP_006715305.1 | 2 - 3 | 435 | 62 | ||
| XM_017011462.1 | XP_016866951.1 | 2 - 3 | 286 | 62 | ||
| XM_017011463.1 | XP_016866952.1 | 2 - 3 | 291 | 62 | ||
| GenBank mRNA | AB014546.1 | - | 2 - 3 | 354 | 62 | |
| AF334675.1 | - | 2 - 3 | 242 | 62 | ||
| AK022075.1 | - | 2 - 3 | 336 | 62 | ||
| AK222765.1 | - | 2 - 3 | 353 | 62 | ||
| AK308840.1 | - | 2 - 3 | 342 | 62 | ||
| AL711279.1 | - | 2 - 3 | 393 | 62 | ||
| BC007517.2 | - | 2 - 3 | 364 | 3028746 | 62 | |
| BT007446.1 | - | 2 - 3 | 242 | 62 | ||
| DA064222.1 | - | 2 - 3 | 435 | 62 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map