Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183078.2 | NP_898901.1 | 6 - 7 | 1431 | 84 | |
| XM_006715242.2 | XP_006715305.1 | 5 - 6 | 1341 | 84 | ||
| XM_017011463.1 | XP_016866952.1 | 6 - 7 | 1287 | 84 | ||
| XM_017011464.1 | XP_016866953.1 | 6 - 7 | 1436 | 84 | ||
| GenBank mRNA | BG029939.1 | - | 4 - 5 | 464 | 84 | |
| BM979512.1 | - | 1 - 2 | 504 | 84 | ||
| CA418096.1 | - | 1 - 2 | 507 | 84 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map