Assay Details
| Gene Symbol: OVCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183378.2 | NP_899234.2 | 27 - 28 | 3332 | 103 | |
| XM_017019257.1 | XP_016874746.1 | 27 - 28 | 5444 | 103 | ||
More Information
Set Membership: |
|

OVCH1
Genomic Map