Assay Details
| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207320.2 | NP_997203.1 | 1 | 818 | 70 | |
| GenBank mRNA | AK098697.1 | - | 1 | 818 | 70 | |
| BC137355.1 | - | 1 | 814 | 9021353 | 70 | |
| BC137356.1 | - | 1 | 814 | 9021354 | 70 | |
More Information
Set Membership: |
|

OTUD6A
Genomic Map