Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146172.1 | NP_001139644.1 | 1 - 2 | 165 | 85 | |
| NM_001146173.1 | NP_001139645.1 | 1 - 2 | 165 | 85 | ||
| NM_033438.3 | NP_254273.2 | 1 - 2 | 165 | 85 | ||
| GenBank mRNA | AF275725.1 | - | 1 - 2 | 132 | 85 | |
| AY034613.1 | - | 1 - 2 | 165 | 85 | ||
| AY358801.1 | - | 1 - 2 | 128 | 85 | ||
| BC070213.1 | - | 1 - 2 | 93 | 85 | ||
| BC074754.2 | - | 1 - 2 | 83 | 30915213 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF275725.1, AY034613.1, AY358801.1, BC070213.1, BC074754.2, NM_001146172.1, NM_001146173.1, NM_033438.3 transcripts.
Set Membership: |
|

SLAMF9
Genomic Map