Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146172.1 | NP_001139644.1 | 2 - 3 | 506 | 73 | |
| GenBank mRNA | AY358801.1 | - | 2 - 3 | 73 | ||
More Information
Set Membership: |
|

SLAMF9
Genomic Map