Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146173.1 | NP_001139645.1 | 2 - 3 | 372 | 68 | |
| GenBank mRNA | BC070213.1 | - | 2 - 3 | 300 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070213.1 transcripts.
Set Membership: |
|

SLAMF9
Genomic Map