Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146319.1 | NP_001139791.1 | 12 - 13 | 1438 | 93 | |
| NM_001146320.1 | NP_001139792.1 | 11 - 12 | 1475 | 93 | ||
| NM_001146321.1 | NP_001139793.1 | 13 - 14 | 1630 | 93 | ||
| NM_001146322.1 | NP_001139794.1 | 12 - 13 | 1249 | 93 | ||
| NM_023927.2 | NP_076416.2 | 12 - 13 | 1587 | 93 | ||
| XM_005272057.3 | XP_005272114.1 | 12 - 13 | 1328 | 93 | ||
| XM_005272058.3 | XP_005272115.1 | 12 - 13 | 1325 | 93 | ||
| XM_005272059.4 | XP_005272116.1 | 13 - 14 | 1718 | 93 | ||
| XM_005272060.3 | XP_005272117.1 | 12 - 13 | 1415 | 93 | ||
| XM_005272061.3 | XP_005272118.1 | 12 - 13 | 1410 | 93 | ||
| XM_005272062.3 | XP_005272119.1 | 12 - 13 | 1376 | 93 | ||
| XM_005272063.3 | XP_005272120.1 | 12 - 13 | 1522 | 93 | ||
| XM_005272064.3 | XP_005272121.1 | 13 - 14 | 1435 | 93 | ||
| XM_011543591.2 | XP_011541893.1 | 13 - 14 | 2306 | 93 | ||
| XM_011543592.2 | XP_011541894.1 | 13 - 14 | 2299 | 93 | ||
| XM_011543593.2 | XP_011541895.1 | 12 - 13 | 1415 | 93 | ||
| XM_011543594.2 | XP_011541896.1 | 12 - 13 | 1187 | 93 | ||
| XM_011543595.2 | XP_011541897.1 | 11 - 12 | 1182 | 93 | ||
| XM_017009781.1 | XP_016865270.1 | 13 - 14 | 1193 | 93 | ||
| XM_017009782.1 | XP_016865271.1 | 13 - 14 | 1432 | 93 | ||
| XM_017009783.1 | XP_016865272.1 | 11 - 12 | 1043 | 93 | ||
| XM_017009784.1 | XP_016865273.1 | 11 - 12 | 1234 | 93 | ||
| XM_017009785.1 | XP_016865274.1 | 11 - 12 | 1284 | 93 | ||
| GenBank mRNA | AK024966.1 | - | 12 - 13 | 1355 | 93 | |
| AK225829.1 | - | 12 - 13 | 1355 | 93 | ||
| AK293307.1 | - | 11 - 12 | 1180 | 93 | ||
| AK296267.1 | - | 13 - 14 | 1335 | 93 | ||
| AK298349.1 | - | 12 - 13 | 1115 | 93 | ||
| AK300154.1 | - | 12 - 13 | 1383 | 93 | ||
| AK303857.1 | - | 12 - 13 | 1189 | 93 | ||
| AK316288.1 | - | 12 - 13 | 1402 | 93 | ||
| AY116970.1 | - | 12 - 13 | 1157 | 93 | ||
| BC008590.1 | - | 12 - 13 | 1320 | 4148772 | 93 | |
More Information
Set Membership: |
|

GRAMD3
Genomic Map