Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146319.1 | NP_001139791.1 | 1 - 2 | 370 | 115 | |
| XM_011543593.2 | XP_011541895.1 | 1 - 2 | 344 | 115 | ||
| XM_017009781.1 | XP_016865270.1 | 1 - 2 | 79 | 115 | ||
| GenBank mRNA | AK300154.1 | - | 1 - 2 | 315 | 115 | |
| DC424558.1 | - | 1 - 2 | 370 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009781.1 transcripts.
Set Membership: |
|

GRAMD3
Genomic Map