Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146322.1 | NP_001139794.1 | 1 - 2 | 121 | 89 | |
| XM_011543594.2 | XP_011541896.1 | 1 - 2 | 56 | 89 | ||
| GenBank mRNA | AK303857.1 | - | 1 - 2 | 61 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK303857.1, NM_001146322.1, XM_011543594.2 transcripts.
Set Membership: |
|

GRAMD3
Genomic Map