Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164803.1 | NP_001158275.1 | 6 - 7 | 748 | 100 | |
| GenBank mRNA | AK304868.1 | - | 6 - 7 | 693 | 100 | |
More Information
Set Membership: |
|

RBMX
Genomic Map