Assay Details
| Gene Symbol: CFHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166624.1 | NP_001160096.1 | 3 - 4 | 522 | 83 | |
| GenBank mRNA | AK298459.1 | - | 3 - 4 | 506 | 83 | |
More Information
Set Membership: |
|

CFHR3
Genomic Map