Assay Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171864.1 | NP_001165335.1 | 3 - 4 | 312 | 83 | |
| NM_024803.2 | NP_079079.1 | 3 - 4 | 432 | 83 | ||
| GenBank mRNA | AK025318.1 | - | 3 - 4 | 434 | 83 | |
| BC098247.1 | - | 3 - 4 | 305 | 40005930 | 83 | |
| BC099716.1 | - | 3 - 4 | 425 | 40005934 | 83 | |
| BC105634.1 | - | 3 - 4 | 425 | 40005931 | 83 | |
| DA890426.1 | - | 3 - 4 | 437 | 83 | ||
More Information
Set Membership: |
|

TUBAL3
Genomic Map