Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184872.1 | NP_001171801.1 | 1 - 2 | 367 | 81 | |
| GenBank mRNA | AF329491.1 | - | 1 - 2 | 130 | 81 | |
More Information
Set Membership: |
|

FCRLA
Genomic Map