Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190328.1 | NP_001177257.1 | 3 - 4 | 348 | 99 | |
| NM_198537.3 | NP_940939.2 | 4 - 5 | 498 | 99 | ||
| XM_005259903.4 | XP_005259960.1 | 3 - 4 | 830 | 99 | ||
| XM_011527997.1 | XP_011526299.1 | 4 - 5 | 764 | 99 | ||
| XM_011527998.2 | XP_011526300.1 | 4 - 5 | 1044 | 99 | ||
| XM_011527999.1 | XP_011526301.1 | 4 - 5 | 573 | 99 | ||
| XM_017026770.1 | XP_016882259.1 | 3 - 4 | 546 | 99 | ||
| XM_017026771.1 | XP_016882260.1 | 3 - 4 | 1127 | 99 | ||
| GenBank mRNA | AK094976.1 | - | 1 - 2 | 1461 | 99 | |
| AK126916.1 | - | 3 - 4 | 2237 | 99 | ||
| BC093948.1 | - | 1 - 2 | 428 | 99 | ||
| BC112285.1 | - | 1 - 2 | 428 | 99 | ||
| BI520231.1 | - | 3 - 4 | 349 | 99 | ||
| DQ778079.1 | - | 3 - 4 | 276 | 99 | ||
| DQ778080.1 | - | 4 - 5 | 426 | 99 | ||
More Information
Set Membership: |
|

YJEFN3
Genomic Map