Assay Details
| Gene Symbol: GFY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195256.1 | NP_001182185.1 | 2 - 3 | 1361 | 98 | |
| XM_011526299.2 | XP_011524601.1 | 3 - 4 | 1473 | 98 | ||
| GenBank mRNA | DN930573.1 | - | 2 - 3 | 216 | 98 | |
More Information
Set Membership: |
|

GFY
Genomic Map