Assay Details
| Gene Symbol: CMTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199317.1 | NP_001186246.1 | 1 - 2 | 445 | 58 | |
| GenBank mRNA | AY862139.1 | - | 1 - 2 | 445 | 58 | |
More Information
Set Membership: |
|

CMTM2
Genomic Map