Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199329.2 | NP_001186258.2 | 3 - 4 | 406 | 115 | |
| GenBank mRNA | AK304490.1 | - | 3 - 4 | 374 | 115 | |
| KU178440.1 | - | 3 - 4 | 339 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304490.1 transcripts.
Set Membership: |
|

FPGT
Genomic Map