Assay Details
| Gene Symbol: RPL17-C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199355.1 | NP_001186284.1 | 5 - 6 | 524 | 121 | |
| NM_001199356.1 | NP_001186285.1 | 5 - 6 | 591 | 121 | ||
| GenBank mRNA | BI461805.1 | - | 5 - 6 | 511 | 122 | |
| BX393840.2 | - | 5 - 6 | 526 | 121 | ||
More Information
Set Membership: |
|

RPL17-C18orf32
Genomic Map