Assay Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204212.1 | NP_001191141.1 | 2 - 3 | 814 | 104 | |
| GenBank mRNA | BC042066.1 | - | 2 - 3 | 814 | 104 | |
More Information
Set Membership: |
|

TMEM235
Genomic Map