Assay Details
| Gene Symbol: KCNJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260508.1 | NP_001247437.1 | 1 | 218 | 72 | |
| NM_001260509.1 | NP_001247438.1 | 1 | 218 | 72 | ||
| NM_001260510.1 | NP_001247439.1 | 1 | 218 | 72 | ||
| NM_002239.3 | NP_002230.1 | 1 | 218 | 72 | ||
| GenBank mRNA | AK293824.1 | - | 1 | 96 | 72 | |
| BC022495.1 | - | 1 | 53 | 4817322 | 72 | |
| GU068048.1 | - | 1 | 47 | 72 | ||
| GU074515.1 | - | 1 | 47 | 72 | ||
| GU074516.1 | - | 1 | 47 | 72 | ||
| U50964.1 | - | 1 | 219 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293824.1, BC022495.1, GU068048.1, GU074515.1, GU074516.1, NM_001260508.1, NM_001260509.1, NM_001260510.1, NM_002239.3, U50964.1 transcripts.
Set Membership: |
|

KCNJ3
Genomic Map