Assay Details
| Gene Symbol: FGFR1OP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278690.1 | NP_001265619.1 | 4 - 5 | 400 | 74 | |
| NM_007045.3 | NP_008976.1 | 4 - 5 | 400 | 74 | ||
| NM_194429.2 | NP_919410.1 | 4 - 5 | 400 | 74 | ||
| GenBank mRNA | AJ420472.1 | - | 4 - 5 | 296 | 74 | |
| AK289846.1 | - | 4 - 5 | 323 | 74 | ||
| AK294950.1 | - | 4 - 5 | 323 | 74 | ||
| AK312791.1 | - | 4 - 5 | 311 | 74 | ||
| BC011902.2 | - | 4 - 5 | 313 | 4120239 | 74 | |
| DC418208.1 | - | 4 - 5 | 401 | 74 | ||
| Y18046.1 | - | 4 - 5 | 389 | 74 | ||
More Information
Set Membership: |
|

FGFR1OP
Genomic Map