Assay Details
| Gene Symbol: AKR1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253909.1 | NP_001240838.1 | 1 - 2 | 150 | 96 | |
| NM_003739.5 | NP_003730.4 | 1 - 2 | 150 | 96 | ||
| GenBank mRNA | AB018580.1 | - | 1 - 2 | 128 | 96 | |
| AK290365.1 | - | 1 - 2 | 110 | 96 | ||
| AK296701.1 | - | 1 - 2 | 112 | 96 | ||
| AK310940.1 | - | 1 - 2 | 108 | 96 | ||
| AK315411.1 | - | 1 - 2 | 110 | 96 | ||
| AV761799.2 | - | 1 - 2 | 152 | 96 | ||
| BC001479.2 | - | 1 - 2 | 2988160 | 96 | ||
| BC019230.1 | - | 1 - 2 | 107 | 3682448 | 96 | |
| BG547635.1 | - | 1 - 2 | 151 | 96 | ||
| BT007286.1 | - | 1 - 2 | 96 | |||
| D17793.1 | - | 1 - 2 | 132 | 96 | ||
| S68288.1 | - | 1 - 2 | 91 | 96 | ||
More Information
Set Membership: |
|

AKR1C3
Genomic Map