Assay Details
| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321111.1 | NP_001308040.1 | 2 - 3 | 220 | 122 | |
| GenBank mRNA | BC095429.1 | - | 2 - 3 | 141 | 122 | |
More Information
Set Membership: |
|

RPS16
Genomic Map