Assay Details
| Gene Symbol: MICB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289160.1 | NP_001276089.1 | 5 | 1027 | 72 | |
| NM_001289161.1 | NP_001276090.1 | 5 | 997 | 72 | ||
| NM_005931.4 | NP_005922.2 | 5 | 1126 | 72 | ||
| GenBank mRNA | AF021221.1 | - | 4 | 912 | 72 | |
| AF021222.1 | - | 4 | 912 | 72 | ||
| AF021223.1 | - | 4 | 912 | 72 | ||
| AF021224.1 | - | 4 | 912 | 72 | ||
| AF021225.1 | - | 4 | 912 | 72 | ||
| AF021226.1 | - | 4 | 912 | 72 | ||
| AK291927.1 | - | 5 | 1124 | 72 | ||
| AK300791.1 | - | 5 | 1027 | 72 | ||
| AK303655.1 | - | 4 | 1261 | 72 | ||
| AK314228.1 | - | 5 | 1031 | 72 | ||
| AY885251.1 | - | 6 | 1078 | 72 | ||
| BC044218.1 | - | 5 | 961 | 6025253 | 72 | |
| U95729.1 | - | 4 | 913 | 72 | ||
| U95730.1 | - | 4 | 913 | 72 | ||
| U95731.1 | - | 4 | 913 | 72 | ||
| U95732.1 | - | 4 | 913 | 72 | ||
| U95733.1 | - | 4 | 913 | 72 | ||
| U95734.1 | - | 4 | 913 | 72 | ||
| X91625.1 | - | 5 | 987 | 72 | ||
More Information
Set Membership: |
|

MICB
Genomic Map