Assay Details
| Gene Symbol: CCL4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291469.1 | NP_001278398.1 | 3 | 417 | 91 | |
| NM_001291470.1 | NP_001278399.1 | 2 | 819 | 91 | ||
| NM_001291471.1 | NP_001278400.1 | 3 | 397 | 91 | ||
| NM_001291472.1 | NP_001278401.1 | 3 | 376 | 91 | ||
| NM_001291473.1 | NP_001278402.1 | 3 | 346 | 91 | ||
| NM_001291474.1 | NP_001278403.1 | 2 | 302 | 91 | ||
| NM_001291475.1 | NP_001278404.1 | 3 | 407 | 91 | ||
| GenBank mRNA | AY766451.1 | - | 3 | 417 | 91 | |
| AY766452.1 | - | 2 | 819 | 91 | ||
| AY766453.1 | - | 3 | 397 | 91 | ||
| AY766454.1 | - | 3 | 376 | 91 | ||
| AY766455.1 | - | 3 | 346 | 91 | ||
| AY766457.1 | - | 2 | 302 | 91 | ||
| AY766458.1 | - | 3 | 407 | 91 | ||
| BC092445.1 | - | 3 | 414 | 30416435 | 91 | |
| CD366524.1 | - | 1 | 450 | 91 | ||
| CD521054.1 | - | 1 | 146 | 91 | ||
More Information
Set Membership: |
|

CCL4L2
Genomic Map