Assay Details
| Gene Symbol: ZNF503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032772.5 | NP_116161.2 | 2 | 2812 | 104 | |
| GenBank mRNA | AK092897.1 | - | 1 | 1931 | 104 | |
| BC011625.2 | - | 2 | 2585 | 4053098 | 104 | |
| BC013011.2 | - | 3 | 2335 | 3604473 | 104 | |
| BC021165.2 | - | 3 | 2335 | 3830632 | 104 | |
| BQ574961.1 | - | 1 | 542 | 104 | ||
More Information
Set Membership: |
|

ZNF503
Genomic Map