Assay Details
| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198989.3 | NP_945340.2 | 2 | 2616 | 72 | |
| GenBank mRNA | AW290996.1 | - | 1 | 367 | 72 | |
| BM931512.1 | - | 1 | 79 | 72 | ||
More Information
Set Membership: |
|

DLEU7
Genomic Map