Assay Details
| Gene Symbol: MIR3683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037454.1 | 1 | 38 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
CCZ1 homolog B, vacuolar protein trafficking and biogenesis associated |
CCZ1B |
C7orf28B, H_NH0577018.2 |
-240.7kb | Sense | Chr. 7 - 6798935 - 6826295 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 655 pseudogene | LOC100131257 | 8.7kb | Sense | Chr. 7 - 7075770 - 7096786 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR3683
Genomic Map