Assay Details
| Gene Symbol: MIR539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030256.1 | 1 | 33 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 487b | MIR487B |
MIRN487B, mir-487b |
-783.0 | Sense | Chr. 14 - 101046455 - 101046538 on NCBI Build GRCh38 |
| Downstream | microRNA 889 | MIR889 |
MIRN889, hsa-mir-889, mir-889 |
503.0 | Sense | Chr. 14 - 101047901 - 101047979 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR539
Genomic Map