| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009402.hg.1 |
ENST00000444970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009402.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009402.hg.1 |
TC09000741.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009401.hg.1 |
ENST00000444970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009401.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009401.hg.1 |
TC09000741.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009399.hg.1 |
ENST00000444970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009399.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009399.hg.1 |
TC09000741.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009400.hg.1 |
ENST00000444970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009400.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009400.hg.1 |
TC09000741.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900160974.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160974.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160974.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160974.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160974.hg.1 |
TC0900008969.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
ENST00000393293.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
ENST00000393293.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160977.hg.1 |
TC0900008969.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
ENST00000393293.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
ENST00000393293.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160976.hg.1 |
TC0900008969.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
ENST00000393293.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
ENST00000393293.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160978.hg.1 |
TC0900008969.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160973.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160973.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160973.hg.1 |
TC0900008969.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
ENST00000318560.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
NM_005157.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
ENST00000318560.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
ENST00000372348.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
NM_005157.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
NM_007313.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160990.hg.1 |
TC0900008969.hg.1 |
Clariom D TC |